Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Calretinin Ab-1 Mouse Monoclonal Antibody, supernatant

Catalog No. MS1110S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1110S0 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1110S0 Supplier Epredia™ Supplier No. MS1110S0

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ Calretinin Ab-1, Mouse Monoclonal Antibody.

Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.

Host Species: Mouse

Clone: 5A5

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length mouse calretinin protein

Molecular Weight: 31.5 kDa

Positive Control: Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting (Not recommended)

Specifications

Antigen Calretinin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5A5
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant full length mouse calretinin protein
Quantity 100 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.