Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calretinin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$440.00
Specifications
Antigen | Calretinin |
---|---|
Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Immunohistochemistry Free-Floating, Knockdown Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Calretinin Polyclonal antibody specifically detects Calretinin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry Free-Floating, KnockDownSpecifications
Calretinin | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating), KnockDown | |
Unconjugated | |
Rabbit | |
Cellular Markers | |
PBS (pH 7.2) and 40% Glycerol | |
794 | |
IgG | |
Immunogen affinity purified |
Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Immunohistochemistry Free-Floating, Knockdown Validated | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LKGFLSDLLKKANRPYDEPKLQEYTQTILRMFDLNGDGKLGLSEMSRLLPVQENFLLKFQGMKLTSEEFNAIFTFYDKDRSGYIDEHELDALLKDLYEKNKKEMNIQQLTNYRKSVMSLAEAGKLYRKDLEIV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title