Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calretinin Antibody (rCALB2/7123), Novus Biologicals™

Mouse Monoclonal Antibody
$237.00 - $474.50
Specifications
Antigen | Calretinin |
---|---|
Clone | rCALB2/7123 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Calretinin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cellular Markers, Phospho Specific | |
10mM PBS with 0.05% BSA | |
794 | |
IgG1 κ | |
Protein A purified |
rCALB2/7123 | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
Recombinant human Calretinin protein fragment (around aa23-242) (exact sequence is proprietary) | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title