Learn More
Invitrogen™ Calretinin Monoclonal Antibody (3G9)

Description
Calretinin Monoclonal Antibody for ICC/IF, IHC (PFA), Western Blot
Specifications
Specifications
| Antigen | Calretinin |
| Applications | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 5mM sodium azide |
| Gene | CALB2 |
| Gene Accession No. | P22676, P47728, Q08331 |
| Gene Alias | 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.