Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calretinin Monoclonal Antibody (6A9)

Catalog No. PIMA547358
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547358 500 μL
PIMA547457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA547358 Supplier Invitrogen™ Supplier No. MA547358
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calretinin Monoclonal Antibody for ICC/IF, IHC (PFA), Western Blot

Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calretinin
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative
Gene CALB2
Gene Accession No. P22676, P47728, Q08331, Q3ZBY3
Gene Alias 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8
Gene Symbols CALB2
Host Species Mouse
Immunogen Full-length recombinant human protein expressed in and purified from E. coli.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100068401, 100127479, 117059, 12308, 513947, 794
Target Species Bovine, Horse, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgA
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.