Learn More
Invitrogen™ Calretinin Monoclonal Antibody (SP13)
Description
This antibody is predicted to react with bovine, chicken and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is mesothelioma.
Specifications
Specifications
| Antigen | Calretinin |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | SP13 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.1% sodium azide; pH 7.20 |
| Gene | CALB2 |
| Gene Accession No. | P22676, P47728, Q08331 |
| Gene Alias | 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.