Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calretinin Mouse anti-Human, Biotin, Clone: 5C5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259473B
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Calretinin | |
Monoclonal | |
Biotin | |
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
Recombinant human Calretinin (1-271aa) purified from E. coli | |
0.1 mL | |
Cellular Markers | |
794 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
5C5 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction