Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calretinin Mouse anti-Human, Clone: 5C5, Novus Biologicals™

Mouse Monoclonal Antibody
$305.00 - $464.00
Specifications
Antigen | Calretinin |
---|---|
Clone | 5C5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1073, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25947350
![]() |
Novus Biologicals
NBP25947350UL |
50 μL |
Each for $305.00
|
|
|||||
NBP259473X
![]() |
Novus Biologicals
NBP259473100UL |
100 μL |
Each for $464.00
|
|
|||||
Description
Calretinin Monoclonal specifically detects Calretinin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Calretinin | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cellular Markers | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
794 | |
Recombinant human CALB2 (1-271aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5C5 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1073, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
CALB2 | |
IgG1 κ | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title