Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calretinin, Rabbit anti-Rat, Human, Polyclonal, Millipore Sigma™

Catalog No. ABN219125UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN219125UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN219125UL Supplier MilliporeSigma Supplier No. ABN219125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Calretinin Antibody, Cat. No. ABN2191, is a highly specific rabbit polyclonal antibody that targets Calretinin and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.

Calretinin (UniProt: P47728; also known as CR) is encoded by the Calb2 gene (Gene ID: 117059) in rat. Calretinin belongs to the calbindin protein family and acts as a calcium-binding protein. It is expressed in both central and peripheral nervous system and is found abundantly in auditory neurons. It is involved in regulating cytosolic calcium ion concentrations and has multiple calcium-binding domains known as EF-hands. Its expression is shown to be regulated at the level of transcription by nuclear respiratory factor 1 (NRF-1) and E2F2 transcription factors. In rat striatum calretinin neurons form a distinct subset of medium-sized (7 20μm), aspiny, GABAergic interneurons. Calretinin can also serve as an important differential diagnostic tool for cystic and solid ameloblastic tumors. (Ref.: Petryszyn, S., et al. (2017). J. Neural Transm. doi: 10.1007/s00702-017-1687; Kresoja-Rakic, J., et al. (2017). Front. Genet. doi: 10.3389/fgene.2017.00070).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calretinin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A 1:500 dilution from a representative lot detected Calretinin in rat brain tissue lysate.Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Calretinin in human cerebral cortex and rat cerebellum tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias CR
Gene Symbols Calb2
Host Species Rabbit
Immunogen GST/His-tagged recombinant fragment corresponding to 99 amino acids from the internal region of rat Calretinin.
Molecular Weight of Antigen ∽31 kDa observed; 31.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Calretinin in human and rat cells. It targets an epitope within 99 amino acids from the internal region.
Target Species Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.