Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calsenilin Antibody (011), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290302UV
Description
Calsenilin Monoclonal specifically detects Calsenilin in Human samples. It is validated for ELISA.Specifications
Calsenilin | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
11 | |
ELISA | |
calsenilin, calsenilin, presenilin-binding protein, EF hand transcription factor, CSEN MGC18289, DREAM, DREAMA-type potassium channel modulatory protein 3, KChIP3, KCHIP3 DRE-antagonist modulator, Kv channel interacting protein 3, calsenilin, Kv channel-interacting protein 3, potassium channel interacting protein 3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Calsenilin (Q9Y2W7-1; Met1-Ile256). | |
0.1 mL | |
Vision | |
30818 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction