Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calsenilin Antibody (011), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290302MFV500
Description
Calsenilin Monoclonal antibody specifically detects Calsenilin in Human samples. It is validated for ELISASpecifications
Calsenilin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
011 | |
ELISA | |
calsenilin, calsenilin, presenilin-binding protein, EF hand transcription factor, CSEN MGC18289, DREAM, DREAMA-type potassium channel modulatory protein 3, KChIP3, KCHIP3 DRE-antagonist modulator, Kv channel interacting protein 3, calsenilin, Kv channel-interacting protein 3, potassium channel interacting protein 3 | |
Rabbit immunized with purified, recombinant Human Calsenilin (Uniprot#: Q9Y2W7-1, Met1-Ile256) | |
0.1 mL | |
Vision | |
30818 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction