Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calsequestrin 2 Antibody (4E10), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174043V
Description
Calsequestrin 2 Monoclonal specifically detects Calsequestrin 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Calsequestrin 2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CASQ2 | |
Recombinant human CASQ2 (20-399aa) purified from E. coli | |
100 μL | |
Cardiovascular Biology, Signal Transduction | |
845 | |
Human, Mouse | |
Purified |
Western Blot, ELISA | |
40000000000 | |
Western Blot, ELISA | |
calsequestrin 2 (cardiac muscle), calsequestrin 2, fast-twitch, cardiac muscle, Calsequestrin, cardiac muscle isoform, calsequestrin-2, FLJ26321, FLJ93514, PDIB2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human CASQ2 mAb, clone 4E10, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CASQ2 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction