Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calsequestrin 2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1C10A1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | Calsequestrin 2 |
|---|---|
| Clone | 1C10A1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6846
![]()
|
Proteintech Group Inc
664191IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Calsequestrin (CASQ) is a Ca2+-binding protein present primarily in junctional sarcoplasmic reticulum of skeletal and cardiac muscle; the cardiac form (CASQ2) is encoded by a separate gene. The primary role of CASQ2 is buffering of the sarcoplasmic reticulum Ca2+ ions, but another role for CASQ2 has emerged recently: CASQ2 regulates the open probability of ryanodine receptor 2 (RyR2). Mutations in CASQ2 cause stress-induced polymorphic ventricular tachycardia, also referred to as catecholaminergic polymorphic ventricular tachycardia 2 (CPVT2), a disease characterized by bidirectional ventricular tachycardia that may lead to cardiac arrest.Specifications
| Calsequestrin 2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Calsequestrin 2, CASQ2, PDIB2 | |
| Casq2 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1C10A1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Pig, Rat | |
| O09161, O14958, O18934, P51868 | |
| 12373, 29209, 397436, 845 | |
| Calsequestrin 2 Fusion Protein Ag13246 | |
| Primary | |
| -20°C | |
| CASQ2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title