Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calsequestrin 2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521706

Catalog No. PIPA521706


Only null left
Add to Cart

Description

Description

Recommended positive controls: HCT116. Predicted reactivity: Mouse (95%), Rat (96%), Dog (96%), Rabbit (96%), Chicken (90%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Calsequestrin (CASQ) is a Ca2+-binding protein present primarily in junctional sarcoplasmic reticulum of skeletal and cardiac muscle; the cardiac form (CASQ2) is encoded by a separate gene. The primary role of CASQ2 is buffering of the sarcoplasmic reticulum Ca2+ ions, but another role for CASQ2 has emerged recently: CASQ2 regulates the open probability of ryanodine receptor 2 (RyR2). Mutations in CASQ2 cause stress-induced polymorphic ventricular tachycardia, also referred to as catecholaminergic polymorphic ventricular tachycardia 2 (CPVT2), a disease characterized by bidirectional ventricular tachycardia that may lead to cardiac arrest.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calsequestrin 2
Polyclonal
Unconjugated
Casq2
AA033488; AW146219; calsequestrin; calsequestrin 2; calsequestrin 2 (cardiac muscle); calsequestrin 2, fast-twitch, cardiac muscle; calsequestrin, cardiac muscle isoform; Calsequestrin-2; cardCSQ; cardiac calsequestrin; CASQ2; cCSQ; Csq2; ESTM52; LOW QUALITY PROTEIN: calsequestrin-2; PDIB2; SR calcium binding protein
Rabbit
Antigen affinity chromatography
RUO
12373, 845
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.68 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O09161, O14958
Casq2
Recombinant protein encompassing a sequence within the center region of human Calsequestrin 2. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.