Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calsequestrin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555568 Supplier Invitrogen™ Supplier No. PA555568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEGLNFPTYD GKDRVVSLSE KNFKQVLKKY DLLCLYYHEP VSSDKVTQKQ FQLKEIVLEL VAQVL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

Calsequestrin (CASQ) is a Ca2+-binding protein present primarily in junctional sarcoplasmic reticulum of skeletal and cardiac muscle; the cardiac form (CASQ2) is encoded by a separate gene. The primary role of CASQ2 is buffering of the sarcoplasmic reticulum Ca2+ ions, but another role for CASQ2 has emerged recently: CASQ2 regulates the open probability of ryanodine receptor 2 (RyR2). Mutations in CASQ2 cause stress-induced polymorphic ventricular tachycardia, also referred to as catecholaminergic polymorphic ventricular tachycardia 2 (CPVT2), a disease characterized by bidirectional ventricular tachycardia that may lead to cardiac arrest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calsequestrin 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Casq2
Gene Accession No. O14958
Gene Alias AA033488; AW146219; calsequestrin; calsequestrin 2; calsequestrin 2 (cardiac muscle); calsequestrin 2, fast-twitch, cardiac muscle; calsequestrin, cardiac muscle isoform; Calsequestrin-2; cardCSQ; cardiac calsequestrin; CASQ2; cCSQ; Csq2; ESTM52; LOW QUALITY PROTEIN: calsequestrin-2; PDIB2; SR calcium binding protein
Gene Symbols Casq2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Calsequestrin 2. Recombinant protein control fragment (Product #RP-94140).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 845
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.