Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calsyntenin-1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297117MFV500
Description
Calsyntenin-1 Polyclonal antibody specifically detects Calsyntenin-1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Calsyntenin-1 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
alcadein alpha 1, Alcadein-alpha, ALC-ALPHA, alcalpha1, alcalpha2, Alzheimer-related cadherin-like protein, cadherin-related family member 12, calsyntenin 1, calsyntenin-1, CDHR12, CS1, CSTN1, FLJ32258, KIAA0911alcadein-alpha, Non-classical cadherin XB31alpha, non-classical cadherin XB31alpha1, PIK3CD, XB31alpha | |
Produced in rabbits immunized with purified, recombinant Human Calsyntenin-1 (Accession#: NP_001009566.1; Met1-Thr859) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
22883 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction