Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calumenin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588528 Supplier Invitrogen™ Supplier No. PA588528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPTEKKDRVH HEPQLSDKVH NDAQSFDYDH DAFLGAEEAK TFDQLTPEES KERLGKIVSK IDGDKDGFVT VDELKDWIKF AQKRWIYEDV ERQWKGHDLN EDGLVSWEEY KNATYGYVLD DPDPDDGFNY KQMMVRDERR FKMADKDGDL IATKEEFTAF LHPEEYDYMK DIVVQETMED IDKNADGFID LEEYIGDMYS HDGNTDEPEW VKTEREQFVE FRDKNRDGKM DKEETKDWIL PSDYDHAEAE ARHLVYESDQ NKDGKLTKEE IVDKYDLFVG SQATDFGEAL VRHDEF; Positive Samples: H460, Rat liver; Cellular Location: Endoplasmic reticulum lumen, Melanosome, Sarcoplasmic reticulum lumen.

Caluminin is a 315 amino acid Ca2+-binding member of the CREC, EF-hand protein family. Calumenin is a secreted protein t hat contains six Ca2+-binding (EF-hand) motifs and is expressed in the lumen of the endoplasmic reticulum (ER) and Golgi apparatus. In the presence of Ca2+, Calumenin interacts with serum amyloid P component (SAP) and, together, they may play a role in the immunological defense system and participate in amyloidosis, the pathological formation of amyloid deposits in different types of tissues. Calumenin has an inhibitory effect on the vitamin K-dependent g-carboxylation system which converts vitamin K-dependent proteins to Gla-containing proteins. Calumenin may also be involved in the pathophysiology of thrombosis and/or wound healing by acting in an autocrine or paracrine manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calumenin
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Calu
Gene Accession No. O35783, O43852
Gene Alias 9530075H20Rik; CALU; Calumenin; Cbp50; CBP-50; Crocalbin; crocalbin-like protein; IEF SSP 9302; multiple EF-hand protein; Rcn; reticulocalbin
Gene Symbols Calu
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-315 of human CALU (NP_0012101).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64366, 813
Target Species Human, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.