Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587948 Supplier Invitrogen™ Supplier No. PA587948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVKLGCSFSG KPGKDPGDQD GAAMDSVPLI SPLDISQLQP PLPDQVVIKT QTEYQLSSPD QQNFPDLEGQ RLNCSHPEEG RRLPTAR; Positive Samples: Mouse testis; Cellular Location: Cell membrane, Cytoplasmic vesicle membrane, Single-pass membrane protein.

Calcyon is a single transmembrane protein that interacts with D1 dopamine receptors. Dopamine is a neurotransmitter that regulates synaptic transmission involved in learning and memory. D1 receptors, the most abundant dopamine receptor in the central nervous system, appear to modulate the activity of D2 dopamine receptors, mediate various behavioural responses, and regulate neuron growth and differentiation. Calcyon is present in neuronal cell bodies and processes of the cortex and hippocampus, and it is especially abundant in pyramidal neurons. Interaction of Calcyon with D1 receptors results in a release of intracellular calcium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALY
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CALY
Gene Accession No. Q9DCA7
Gene Alias 0710001P07Rik; 1110004A22Rik; Calcyon; calcyon D1 dopamine receptor-interacting protein (CALCYON); calcyon neuron specific vesicular protein; calcyon neuron-specific vesicular protein; calcyon protein; calcyon; D1 dopamine receptor-interacting protein; CALY; D1 dopamine receptor-interacting protein; dopamine receptor D1 interacting protein; Drd1ip; Neuron-specific vesicular protein calcyon; NSG3
Gene Symbols CALY
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-87 of human CALY (NP_0565371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68566
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.