Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calyculin A, Tocris Bioscience™

Catalog No. 1336100u
Click to view available options
Quantity:
100 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Protein phosphatase 1 and 2A inhibitor

Calyculin A is a potent and selective cell-permeable inhibitor of protein phosphatase 1 (IC50 = 0.3 - 0.7 nM) and protein phosphatase 2A (IC50 = 0.5 - 1 nM). Displays > 10,000,000-fold selectivity over PP2B and PP2C.

Note: This product is typically prepared in DMSO.

Chemical Identifiers

CAS 101932-71-2
Molecular Formula C50H81N4O15P
Molecular Weight (g/mol) 1009.185
InChI Key FKAWLXNLHHIHLA-OROLNYDDSA-N
Synonym calyculin a
PubChem CID 131632568
IUPAC Name [(2R,3R,5R,7R,8R,9S)-2-[(1S,3S,4S,5R,6R)-14-cyano-3,5-dihydroxy-1-methoxy-4,6,8,9,13-pentamethyltetradeca-7,9,11,13-tetraenyl]-9-[3-[2-[(2S)-4-[[(2S,3S,4S)-4-(dimethylamino)-2,3-dihydroxy-5-methoxypentanoyl]amino]butan-2-yl]-1,3-oxazol-4-yl]prop-2-enyl]-7
SMILES CC1C(CC2(C(C(C(O2)C(CC(C(C)C(C(C)C=C(C)C(=CC=CC(=CC#N)C)C)O)O)OC)OP(=O)(O)O)(C)C)OC1CC=CC3=COC(=N3)C(C)CCNC(=O)C(C(C(COC)N(C)C)O)O)O

Specifications

Quantity 100 μg
Formula Weight 1009.18
Percent Purity >98%
Chemical Name or Material Calyculin A
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.