Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase I Antibody (010), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290023AF750
Description
CaM Kinase I Monoclonal specifically detects CaM Kinase I in Human samples. It is validated for ELISA.Specifications
CaM Kinase I | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
10 | |
ELISA | |
calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CaM Kinase I (NP_003647.1; Leu2-Leu370). | |
0.1 mL | |
Neuroscience, Protein Kinase, Sensory Systems, Vision | |
8536 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction