Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase I Rabbit anti-Human, Mouse, Rat, Clone: 4P4K9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | CaM Kinase I |
---|---|
Clone | 4P4K9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CaM Kinase I Monoclonal antibody specifically detects CaM Kinase I in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CaM Kinase I | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience, Protein Kinase, Sensory Systems, Vision | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8536 | |
IgG | |
Affinity purified |
4P4K9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CaM Kinase I (Q14012). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title