Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase II alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | CaM Kinase II alpha |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017825
![]() |
Novus Biologicals
NB100818300.025ML |
0.025 mL |
Each for $181.90
|
|
|||||
NB10081830
![]() |
Novus Biologicals
NB10081830 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
CaM Kinase II alpha Polyclonal specifically detects CaM Kinase II alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CaM Kinase II alpha | |
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin | |
Polyclonal | |
Rabbit | |
Wnt Signaling Pathway | |
Q9UQM7 | |
815 | |
IgG | |
Affinity Purified | |
50 kDa |
1.0 mg/mL | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain | |
CAMK2A | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title