Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase II gamma Sheep anti-Human, Polyclonal, R&D Systems™

Catalog No. AF7280SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7280SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7280SP Supplier R&D Systems Supplier No. AF7280SP
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CaM Kinase II gamma Polyclonal antibody AF7280SP specifically detects CaM Kinase II gamma in Human samples. It is validated for Western Blotting

Specifications

Antigen CaM Kinase II gamma
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 1-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13555
Gene Alias calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CAMK, CAMK2G, CAMKG, CAMKGFLJ16043, CAMK-II, CaMK-II subunit gamma, EC 2.7.11, EC 2.7.11.17, MGC26678
Gene Symbols CAMK2G
Host Species Sheep
Immunogen E. coli-derived recombinant human CaM Kinase II gamma Ala448-Gln558 Accession # Q13555
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 818
Test Specificity Detects human CaM Kinase II gamma in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) CaM Kinase II beta is observed, and less than 5% cross-reactivity with rhCaM Kinase II alpha and rhCaM Kinase II delta is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.