Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cambridge Diagnostic Products, Inc.™ Camco Quik Stain™ and Quik Stain™ II
Click to view available options
Type:
Quik Stain
Quik Stain II
Description
- Both are self-buffering and intensity controlled by washing with ordinary distilled water
- CAMCO Quik Stain is a Wright's stain formulation (traditional blue coloration) and CAMCO Quik Stain II is a Wright-Giemsa stain (traditional coloration with blue-red highlights)
Order Info
Packaged in 946mL bottles
Specifications
Specifications
Quantity | 946 mL |
Boiling Point | 64°C |
Format | Liquid |
Type | Quik Stain |
Color | Dark Blue |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction