Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cambridge Diagnostic Products, Inc.™ Camco™ Stain Pak

Catalog No. p-121832
Click to view available options
Quantity:
3 x 473 mL
1 gal.
Type:
Solution I (Red)
Solution II (Blue)
Stain Kit
Stain Pak
This item is not returnable. View return policy
This item is not returnable. View return policy

  • Provides rapid, consistent differential staining of blood smears approximating results obtained from Wright-Giemsa staining
  • In addition to highly purified dyestuff and buffers, CAMCO Stain Pak contains important electrolytes that enhance the staining of the nuclei and improve contrast
  • Fixative solution is used to stabilize the cell and its contents
  • Convenient sizes available

Order Info

Stains are packaged in one-gallon bottles except for Stain Pak, which is packaged in one 473mL bottle each of Fixative, Solution I, and Solution II

Specifications

Quantity 3 x 473 mL
Boiling Point 100°C
Format Liquid
Type Stain Pak
For Use With (Application) For rapid differential staining of Blood smears
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.