Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cambridge Diagnostic Products, Inc.™ Camco™ Wright Stain, Fixative and Buffer

Catalog No. p-121807
Click to view available options
Quantity:
1 gal.
10 x 4.4 g Vial
Type:
Buffer
Fixative and Buffer
Stain
This item is not returnable. View return policy
This item is not returnable. View return policy

Plastic container is easy to store and allows convenient dispensing into slide stainer traps

  • Contains the optimum concentration of stain to give accurate results when used on the Hemtrak™ WBC Differential System and for manual staining
  • Plastic container is easy to store and allows convenient dispensing into slide stainer traps
  • Wright stain buffer is helpful with the development of the correct, clear nuclear color schemes and has a pH of 7.15 (10 × 4.4g vial; each vial prepares 1 gallon of pH 7.15 buffer)
  • Directions for use printed on Wright stain container

Specifications

Quantity 1 gal.
Boiling Point 64°C
Format Liquid
Type Stain
For Use With (Application) For in-vitro diagnostic use
Color Dark Blue
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.