Learn More
Promega CAMK1 gamma Kinase Enzyme System, 1mg
Catalog No. PRV6277 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6277 1 mg
1 options
Catalog No. PRV6277 Supplier Promega Supplier No. V6277
Add to Cart
Add to Cart
Request a Quote
Full-length recombinant human CAMK1 gamma was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK1 gamma (CLICK-III), a member of the CAMK family, is a novel membrane-anchored neuronal Ca2+/calmodulin-dependent protein kinase.
  • Easily Screen and Profile CAMK1γ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C