Learn More
CAMK1D Polyclonal Antibody, Invitrogen™
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (84%), Pig (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | CAMK1D |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.01 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | CAMK1D |
| Gene Accession No. | Q8IU85 |
| Gene Alias | A630059D12Rik; calcium/calmodulin dependent protein kinase ID; calcium/calmodulin-dependent protein kinase 1D; calcium/calmodulin-dependent protein kinase ID; calcium/calmodulin-dependent protein kinase type 1D; CaM kinase I delta; caM kinase ID; CaM-K1; CAMK1D; CAMKI; caMKI delta; caM-KI delta; CAMKID; CaMKIdelta; CaMKI-delta; camKI-like protein kinase; CKLiK; E030025C11Rik; mCKLiK; MGC120317; MGC120318; OTTHUMP00000019119; OTTHUMP00000019120; OTTHUMP00000045030; RGD1560691; RP11-462F15.1 |
| Gene Symbols | CAMK1D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.