Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CAMK1G (CaMKI gamma), GST Tag Recombinant Protein

Catalog No. A30489
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg

Recombinant Protein

CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96NX5
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 57172
Molecular Weight (g/mol) 79.8 kDa
Name Human CAMK1G (CaMKI gamma), GST Tag
Quantity 10 μg
Gene Alias calcium/calmodulin dependent protein kinase IG; calcium/calmodulin-dependent protein kinase I gamma; calcium/calmodulin-dependent protein kinase IG; calcium/calmodulin-dependent protein kinase type 1 G; CaM kinase 1 G; CaM kinase I gamma; caM kinase IG; CAMK1G; caMKI gamma; caM-KI gamma; caMKIG; CaMKIgamma; caMKI-gamma; caMK-like CREB kinase III; CLICK III; CLICK3; CLICKIII; CLICK-III; dJ272L16.1; RP1-272L16.2; VWS1
Common Name CaMKI gamma
KinaseFamily CAMK1 Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.