Learn More
Promega CAMK2 alpha Kinase Enzyme System, 10microg
Catalog No. PRV4018 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4018 10 μg
1 options
Catalog No. PRV4018 Supplier Promega Supplier No. V4018
Recombinant full-length human CAMK2 alpha was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. CAMK2 alpha is a ser/thr protein kinase that is a member of the Ca2+/calmodulin-dependent protein kinase family.
  • Easily Screen and Profile CAMK2α Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C