Learn More
Promega CAMK2 gamma Kinase Enzyme System, 1mg
Catalog No. PRV3532 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3532 1 mg
1 options
Catalog No. PRV3532 Supplier Promega Supplier No. V3532
Add to Cart
Add to Cart
Request a Quote
Recombinant human CAMK2 gamma (C-terminal truncation) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. CAMK2 gamma is a member of the CAMKII family.
  • Easily Screen and Profile CAMK2γ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C