Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camk2a Mouse anti-Rat, APC, Clone:6G9, Abnova™

Catalog No. 89333710 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-710 Supplier Abnova Corporation Supplier No. MAB16954
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against rat Camk2a.

Specifications

Antigen Camk2a
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 6G9
Conjugate APC
Description calcium/calmodulin-dependent protein kinase II alpha
Dilution ELISA
Immunocytochemistry (1:50)
Immunofluorescence (1:50)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:2000)
Immunoprecipitation
Radioimmunoassay
Western Blot (1:10000)
The optimal working dilution should b
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Camk2a
Gene Alias PK2CDD, PKCCD
Host Species Mouse
Immunogen Partially purified rat Camk2a.
Quantity 100 μg
Gene ID (Entrez) 25400
Target Species Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.