Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camk2a, Mouse anti-Rat, ATTO390, Clone: 6G9, Abnova™

Catalog No. 89337959 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-959 Supplier Abnova Corporation Supplier No. MAB18034
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen Camk2a
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 6G9
Conjugate Atto 390
Dilution ELISA Immunocytochemistry (1:50) Immunofluorescence (1:50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:2000) Immunoprecipitation Radioimmunoassay Western Blot (1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Gene calcium/calmodulin-dependent protein kinase II alpha
Gene Alias PK2CDD/PKCCD
Gene Symbols Camk2a
Host Species Mouse
Immunogen Partially purified rat Camk2a.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25400
Target Species Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.