Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camk2b Mouse anti-Human, Mouse, Rat, Clone: CB-beta-1, Abnova™

Catalog No. 89035412 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-412 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-412 Supplier Abnova Corporation Supplier No. MAB14007

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native rat Camk2b.

  • High specificity
  • High reactivity

Specifications

Antigen Camk2b
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CB-beta-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native rat Camk2b.
Dilution ELISA Flow Cytometry (3-5 ug x 106 cells) Immunohistochemistry Immunoprecipitation Western Blot (0.5-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide).
Gene Accession No. P08413
Gene Alias Ck2b
Gene Symbols Camk2b
Host Species Mouse
Immunogen Native purified Camk2b from rat brain.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24245
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.