Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMK2G Mouse anti-Human, Clone:6A4A10, Abnova™

Catalog No. 89333207 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-207 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-207 Supplier Abnova Corporation Supplier No. MAB16592
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human CAMK2G.

The product of this gene belongs to the Serine/Threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells the enzyme is composed of four different chains: alpha, beta, gamma, and delta. The product of this gene is a gamma chain. Six alternatively spliced variants that encode six different isoforms have been characterized to date. Additional alternative splice variants that encode different isoforms have been described, but their full-length nature has not been determined. [provided by RefSeq]

Specifications

Antigen CAMK2G
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6A4A10
Conjugate Unconjugated
Description calcium/calmodulin-dependent protein kinase II gamma
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CAMK2G
Gene Alias CAMK, CAMK-II, CAMKG, FLJ16043, MGC26678
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 322-481 of human CAMK2G from E. coli.
Quantity 100 μg
Gene ID (Entrez) 818
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.