Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAMK2N2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565644 Supplier Invitrogen™ Supplier No. PA565644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILPYSEDKMGR FGADPEGSDL SFSCR Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

Potent and specific cellular inhibitor of CaM-kinase II (CAMK2). Traps Ca(2+)/calmodulin on CAMK2. May play an important role in the regulation of cell growth when overexpressed in colon adenocarcinoma LoVo cells. Traps Ca(2+)/calmodulin on CAMK2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMK2N2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAMK2N2
Gene Accession No. Q96S95
Gene Alias calcium/calmodulin dependent protein kinase II inhibitor 2; calcium/calmodulin-dependent protein kinase II inhibitor 2; CAMK2N2; CaM-KII inhibitory protein; CAMKIIN; CAM-KIIN
Gene Symbols CAMK2N2
Host Species Rabbit
Immunogen Recombinant Human CAMK2N2. Recombinant protein control fragment (Product #RP-106316).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94032
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.