Learn More
Promega CAMK4 Kinase Enzyme System, 1mg
Catalog No. PRV2952 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2952 1 mg
1 options
Catalog No. PRV2952 Supplier Promega Supplier No. V2952
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human CAMK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CAMK4 is a multifunctional serine/threonine protein kinase and a member of Ca2+/calmodulin-dependent protein kinase family.
  • Easily Screen and Profile CAMK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C