Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaMKI Recombinant Rabbit Monoclonal Antibody (010), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543064
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543064 100 μL
PIMA543469 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543064 Supplier Invitrogen™ Supplier No. MA543064
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human CAMKI/CAMK1

CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaMKI
Applications ELISA
Classification Recombinant Monoclonal
Clone 010
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CAMK1
Gene Accession No. Q14012
Gene Alias AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19
Gene Symbols CAMK1
Host Species Rabbit
Immunogen Recombinant Human CAMKI/CAMK1 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8536
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.