Learn More
Invitrogen™ CaMKI Recombinant Rabbit Monoclonal Antibody (4P4K9)

Description
Positive Samples: HepG2, THP-1, Mouse lung, Mouse liver, Mouse brain, Rat lung, Rat liver, Rat brain Immunogen sequence: MLGAVEGPRW KQAEDIRDIY DFRDVLGTGA FSEVILAEDK RTQKLVAIKC IAKEALEGKE GSMENEIAVL HKIKHPNIVA LDDIYESGGH LYLIMQLVSG.
Specifications
Specifications
| Antigen | CaMKI |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 4P4K9 |
| Concentration | 0.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CAMK1 |
| Gene Accession No. | Q14012, Q63450, Q91YS8 |
| Gene Alias | AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.