Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMKID Antibody (OTI2C6), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272074AF594
Description
CAMKID Monoclonal specifically detects CAMKID in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CAMKID | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CAMK1D | |
Full length recombinant protein of human CAMKID (NP_065130) produced in HEK293T cells. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2C6 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
calcium/calmodulin-dependent protein kinase ID, calcium/calmodulin-dependent protein kinase type 1D, CaM kinase I delta, CaM kinase ID, CaM-K1, CaMKI delta, CaM-KI delta, CaMKID, CaMKI-like protein kinase, CKLiKEC 2.7.11.17, EC 2.7.11 | |
Mouse | |
0.1 mL | |
57118 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction