Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaMKII alpha/beta, p Thr287, p Thr286 Antibody (22B1), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11096869G
Description
CaMKII alpha/beta Monoclonal specifically detects CaMKII alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
CaMKII alpha/beta (p Thr286, p Thr287) | |
Monoclonal | |
DyLight 488 | |
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody is specific for a and B subunits of CaMKII only when they are phosphorylated at Thr-286/287 (in B). | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot | |
22B1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting | |
CAMK2A | |
Synthetic peptide | |
0.1 mL | |
Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway | |
815 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction