Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CaMKII beta/gamma Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599600
Description
Antibody detects endogenous levels of total CaMKII beta/gamma.
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be coexpressed in the same tissue type. CaMKIV is stimulated by Ca2+ and CaM but also requires phosphorylation by a CaMK for full activation.
Specifications
CaMKII beta/gamma | |
Polyclonal | |
Unconjugated | |
CAMK2B | |
Ca++/calmodulin-dependent protein kinase II beta subunit; Ca2+/calmodulin-dependent protein kinase II; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; calcium/calmodulin-dependent protein kinase type II subunit beta; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II beta subunit; caM kinase II subunit beta; CaM kinase II subunit gamma; CAM2; CAMK; CaMK II; CAMK2; CAMK2B; Camk2d; Camk2g; CAMKB; CAMKG; CAMK-II; caMK-II subunit beta; caMK-II subunit gamma; CaM-kinase II beta chain; caM-kinase II gamma chain; Ck2b; proline rich calmodulin-dependent protein kinase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12323, 12325, 816, 818 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P28652, Q13554, Q13555, Q923T9 | |
CAMK2B, CAMK2G | |
A synthesized peptide derived from human CAMK2G(Accession Q13555), corresponding to amino acid residues D505-A555. | |
100 μL | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction