Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaMKII beta/gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536195 Supplier Invitrogen™ Supplier No. PA536195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 50-80 kDa. Purity is >95% by SDS-PAGE.

The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be coexpressed in the same tissue type. CaMKIV is stimulated by Ca2+ and CaM but also requires phosphorylation by a CaMK for full activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaMKII beta/gamma
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CAMK2B
Gene Accession No. P08413, P11730, P28652, Q13554, Q13555, Q923T9
Gene Alias Ca++/calmodulin-dependent protein kinase II beta subunit; Ca2+/calmodulin-dependent protein kinase II; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II gamma chain; calcium/calmodulin-dependent protein kinase type II subunit beta; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II beta subunit; caM kinase II subunit beta; CaM kinase II subunit gamma; CAM2; CAMK; CaMK II; CAMK2; CAMK2B; Camk2d; Camk2g; CAMKB; CAMKG; CAMK-II; caMK-II subunit beta; caMK-II subunit gamma; CaM-kinase II beta chain; caM-kinase II gamma chain; Ck2b; proline rich calmodulin-dependent protein kinase
Gene Symbols CAMK2B, CAMK2G
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 601-652of human CaMKII beta.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12323, 12325, 171140, 24245, 816, 818
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.