Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CaMKII beta Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586771
Description
Purity is > 95% by SDS-PAGE.
CAMK2B (CaMKII beta) is a serine/threonine kinase that plays a role in synaptic plasticity, synaptic vesicle mobilization and regulation of gene expression. As a dominant kinase in the central nervous system it may function in long term potentiation and neurotransmitter release. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be co-expressed in the same tissue type. CaMKII alpha is autophosphorylated on Thr 286 upon the binding of the Ca2+/CaM complex to the autoinhibitory domain of CaMKII. This process is called Ca2+/CaM trapping, which is thought to be involved in the synaptic encoding of information.
Specifications
CaMKII beta | |
Polyclonal | |
Unconjugated | |
CAMK2B | |
Ca++/calmodulin-dependent protein kinase II beta subunit; calcium/calmodulin dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase (CaM kinase) II beta; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II, beta; calcium/calmodulin-dependent protein kinase type II beta chain; calcium/calmodulin-dependent protein kinase type II subunit beta; CaM kinase II beta subunit; caM kinase II subunit beta; CAM2; CaMK II; CAMK2; CAMK2B; Camk2d; CAMKB; caMK-II subunit beta; caM-kinase II beta chain; Ck2b; hypothetical protein LOC525416; MGC29528; proline rich calmodulin-dependent protein kinase; ß CaMKII; ßCaMKII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12323, 24245, 816 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
P08413, P28652, Q13554 | |
CAMK2B | |
A synthetic peptide corresponding to residues in Human PI3K p85beta. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction