Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaMKIV Monoclonal Antibody (7B8)
GREENER_CHOICE

Catalog No. PIMA532937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532937 Supplier Invitrogen™ Supplier No. MA532937
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Purity is > 95% by SDS-PAGE.

CAMK4 (CaMKIV) is a serine/threonine protein kinase that phosphoryates CREB, oncoprotein 18, and SRF. It has limited tissue distribution, and has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells. The nuclear localization of this protein is consistent with its role in mediating calcium-dependent gene expression. CaMKIV is particularly abundant in testis, T-cells, and neurons but is also found in other tissues to varying degrees. In neurons, CaMKIV is thought to play an important role in synaptic plasticity via its gene regulatory effects. In T-cells, this protein plays an important role in calcium signaling which could affect the transcription regulatory protein, nuclear factor of activated T-cells (NFAT). CaMKIV is encoded, along with calspermin, by the CaMKIV gene. It has been found that, in testes, CaMKIV is expressed in germ cells and found to be associated with chromatin. The association of CaMKIV with chromatin suggests a potential role in chromatin remodeling during nuclear condensation in spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaMKIV
Applications Western Blot
Classification Monoclonal
Clone 7B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CAMK4
Gene Accession No. Q16566
Gene Alias A430110E23Rik; AI666733; brain Ca(2+)-calmodulin-dependent protein kinase type IV; brain Ca++-calmodulin-dependent protein kinase type IV; Ca2+/calmodulin-dependent protein kinase type IV/Gr; Calcium/calmoduli; calcium/calmodulin dependent protein kinase IV; calcium/calmodulin-dependent protein kinase IV; calcium/calmodulin-dependent protein kinase IV L homeolog; calcium/calmodulin-dependent protein kinase type IV; calcium/calmodulin-dependent protein kinase type IV catalytic chain; Calmodulin-dependent protein kinase IV; calspermin; CAM kinase- GR; CAM kinase IV; CAM kinase-GR; CAMK; CaMK IV; camk4; camk4.L; camk4-A; CAMK-GR; CAMKIV; CaMKIV/Gr; Ccdpk; D18Bwg0362e; EC 2.7.11.17; KCC4; kinase CaMK4; OTTHUMP00000222818; RATCCDPK; xCaM; XELAEV_18008011mg
Gene Symbols CAMK4
Host Species Mouse
Immunogen Recombinant full length Human CAMKIV.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 814
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.