Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaMKK alpha Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142682G
Description
CaMKK alpha Polyclonal antibody specifically detects CaMKK alpha in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
CaMKK alpha | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
calcium/calmodulin-dependent protein kinase kinase 1, calcium/calmodulin-dependent protein kinase kinase 1, alpha, Calcium/calmodulin-dependent protein kinase kinase alpha, CaM-kinase IV kinase, CaM-kinase kinase 1, CaM-kinase kinase alpha, CaM-KK 1, CaMKK alpha, CaM-KK alpha, CAMKK alpha protein, CAMKKACaMKK 1, DKFZp761M0423, EC 2.7.11, EC 2.7.11.17, MGC34095 | |
The immunogen for this product maps to a region between residue 150 and 200 of human calcium/calmodulin-dependent protein kinase kinase 1, alpha using the numbering given in entry NP_757343.1 (GeneID 84254). | |
0.1 mL | |
Protein Kinase | |
84254 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction