Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaMKK alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547635
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547635 Supplier Invitrogen™ Supplier No. PA547635
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

This antibody detects endogenous human and rat CaMKK alpha in Western blots. In Western blots, this antibody does not cross-react with recombinant human CaMKK beta. Reconstitute at 0.2 mg/mL in sterile PBS.

CAMKK1 (CAMKKA) is a serine/threonine protein kinase family member, and belongs to the Ca2+/calmodulin-dependent protein kinase subfamily. The product of CaMKK alpha is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. CAMKK1 functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for CaMKK alpha. Different isoforms of this protein may regulate inducible expression of different cytokine genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaMKK alpha
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CAMKK1
Gene Accession No. P97756, Q8N5S9
Gene Alias AI846603; alpha CaMKK; calcium calmodulin dependent protein kinase kinase alpha; calcium/calmodulin dependent protein kinase kinase 1; calcium/calmodulin-dependent protein kinase kinase 1; calcium/calmodulin-dependent protein kinase kinase 1, alpha; calcium/calmodulin-dependent protein kinase kinase alpha; CaM-kinase IV kinase; caM-kinase kinase 1; caM-kinase kinase alpha; Camkk; caMKK 1; caM-KK 1; caMKK alpha; CaM-KK alpha; CAMKK alpha protein; CAMKK1; CAMKKA; CaMKKalpha
Gene Symbols CAMKK1
Host Species Sheep
Immunogen E. coli-derived recombinant human CaMKK alpha Met1-Ser132.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60341, 84254
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.