Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CaMKK beta Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521929
Description
Recommended positive controls: PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAMKK2 (CaMKK beta) is a serine/threonine kinase involved in neurogenesis, cell differentiation, and signal transduction. It also activates CAMKI and CAMKIV, and plays a role in the development of long term memory.
Specifications
CaMKK beta | |
Polyclonal | |
Unconjugated | |
CAMKK2 | |
6330570N16Rik; AW061083; Ca+/Calmodulin-dependent protein kinase kinase beta (CaM-kinase kinase beta); calcium/calmodulin dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase 2 beta; calcium/calmodulin-dependent protein kinase beta; calcium/calmodulin-dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase kinase 2, beta; calcium/calmodulin-dependent protein kinase kinase beta; caM-kinase kinase 2; CaM-kinase kinase beta; CAMKK; caMKK 2; caM-KK 2; caMKK beta; CaM-KK beta; CAMKK beta protein; CAMKK2; CAMKKB; KIAA0787; MGC15254; mKIAA0787 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.98 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q96RR4 | |
CAMKK2 | |
Synthetic peptide corresponding to a region within amino acids 1 and 498 of CaMKK beta. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction