Learn More
Promega CAMKK1 Kinase Enzyme System, 1mg
Catalog No. PRV6279 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6279 1 mg
1 options
Catalog No. PRV6279 Supplier Promega Supplier No. V6279
Add to Cart
Add to Cart
Request a Quote
CAMKK1 or CAMKK alpha is a Ca2+/calmodulin-dependent protein kinase that activates CAM-kinases I and IV via phosphorylation of their Thr(177) and Thr(196) residues, respectively.
  • Easily Screen and Profile CAMKK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C