Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAMKK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102029 Supplier Invitrogen™ Supplier No. PA5102029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CAMKK2.

CAMKK2 (CaMKK beta) is a serine/threonine kinase involved in neurogenesis, cell differentiation, and signal transduction. It also activates CAMKI and CAMKIV, and plays a role in the development of long term memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMKK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CAMKK2
Gene Accession No. O88831, Q8C078, Q96RR4
Gene Alias 6330570N16Rik; AW061083; Ca+/Calmodulin-dependent protein kinase kinase beta (CaM-kinase kinase beta); calcium/calmodulin dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase 2 beta; calcium/calmodulin-dependent protein kinase beta; calcium/calmodulin-dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase kinase 2, beta; calcium/calmodulin-dependent protein kinase kinase beta; caM-kinase kinase 2; CaM-kinase kinase beta; CAMKK; caMKK 2; caM-KK 2; caMKK beta; CaM-KK beta; CAMKK beta protein; CAMKK2; CAMKKB; KIAA0787; MGC15254; mKIAA0787
Gene Symbols CAMKK2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CAMKK2(Accession Q96RR4), corresponding to amino acid residues I485-R535.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10645, 207565, 83506
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.